Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008996.3 | NP_033022.1 | 6 | 507 | ||
GenBank mRNA | AB232583.1 | - | 6 | 479 | ||
AF226873.1 | - | 6 | 507 | |||
AK034408.1 | - | 6 | 675 | |||
AK129477.1 | - | 6 | 665 | |||
AK151085.1 | - | 6 | 671 | |||
AK152978.1 | - | 6 | 676 | |||
AK159425.1 | - | 6 | 671 | |||
AK166249.1 | - | 6 | 668 | |||
AK167150.1 | - | 6 | 675 | |||
AK212539.1 | - | 2 | 177 | |||
BC002077.1 | - | 6 | 674 | 3592802 | ||
Y00094.1 | - | 6 | 608 |