Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159729.1 | NP_001153201.1 | 3 | 453 | ||
NM_008999.4 | NP_033025.2 | 3 | 565 | |||
GenBank mRNA | AB232618.1 | - | 1 | 130 | ||
AK016493.1 | - | 3 | 465 | |||
AK045454.1 | - | 3 | 324 | |||
AK047068.1 | - | 3 | 566 | |||
AK054079.1 | - | 3 | 551 | |||
AK163909.1 | - | 3 | 574 | |||
AK164057.1 | - | 3 | 552 | |||
BC025578.1 | - | 3 | 460 | 5323566 | ||
M79305.1 | - | 1 | 70 | |||
Z22821.1 | - | 1 | 150 |