Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009003.3 | NP_033029.2 | 4 | 390 | ||
GenBank mRNA | AB232591.1 | - | 4 | 250 | ||
AK136566.1 | - | 4 | 379 | |||
AK145452.1 | - | 4 | 373 | |||
AK161877.1 | - | 4 | 358 | |||
BC138508.1 | - | 4 | 303 | 8861528 | ||
CO806247.1 | - | 406 | ||||
D86563.1 | - | 4 | 364 |