Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009003.3 | NP_033029.2 | 8 | 1366 | ||
GenBank mRNA | AK145452.1 | - | 8 | 1349 | ||
AK161877.1 | - | 8 | 1334 | |||
D86563.1 | - | 8 | 1343 |