Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009009.4 | NP_033035.3 | 2, 3 | 381 | ||
GenBank mRNA | AF332085.1 | - | 1, 2 | 136 | ||
AF332086.1 | - | 1, 2 | 136 | |||
AK004746.1 | - | 2, 3 | 184 | |||
AK128984.1 | - | 2, 3 | 270 | |||
AK139492.1 | - | 1, 2 | 34 | |||
AK145591.1 | - | 2, 3 | 348 | |||
AK163992.1 | - | 242 | ||||
AK168718.1 | - | 2, 3 | 369 | |||
AK168896.1 | - | 2, 3 | 330 | |||
BC043032.1 | - | 1, 2 | 150 | 5696540 | ||
BC129916.1 | - | 1, 2 | 180 | 40111820 | ||
BC129917.1 | - | 1, 2 | 181 | 40111821 | ||
D37790.1 | - | 1, 2 | 68 | |||
X98293.1 | - | 1, 2 | 136 |