Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009012.2 | NP_033038.2 | 5 | 834 | ||
GenBank mRNA | AK018001.1 | - | 5 | 834 | ||
AK037641.1 | - | 5 | 835 | |||
AK144155.1 | - | 5 | 826 | |||
BC058180.1 | - | 5 | 810 | |||
U66887.1 | - | 5 | 812 |