Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009012.2 | NP_033038.2 | 20 | 3335 | ||
GenBank mRNA | U66887.1 | - | 20 | 3313 |