Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPARCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128310.2 | NP_001121782.1 | 5 | 814 | ||
NM_001291976.1 | NP_001278905.1 | 5 | 821 | |||
NM_004684.5 | NP_004675.3 | 4 | 712 | |||
GenBank mRNA | AK298039.1 | - | 4 | 420 | ||
AK300956.1 | - | 4 | 415 | |||
AK316533.1 | - | 5 | 411 | |||
AL833430.1 | - | 4 | 450 | |||
AY513278.1 | - | 4 | 322 | |||
BC033721.1 | - | 4 | 487 | 4939390 | ||
BX647713.1 | - | 5 | 363 | |||
DA283085.1 | - | 4 | 415 | |||
X82157.1 | - | 4 | 439 | |||
X86693.1 | - | 4 | 564 |