Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009023.3 | NP_033049.2 | 3 | 780 | ||
GenBank mRNA | BC111863.1 | - | 3 | 736 | 40046186 | |
J03962.1 | - | 3 | 716 | |||
X15788.1 | - | 3 | 769 |