Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009026.4 | NP_033052.1 | 1 | 328 | ||
GenBank mRNA | AF009246.1 | - | 1 | 332 | ||
AK038932.1 | - | 1 | 331 | |||
BC034166.1 | - | 1 | 298 | 4989312 | ||
CT010376.1 | - | 1 | 191 |