Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080927.2 | NP_001074396.1 | 6 | 810 | ||
NM_001080928.1 | NP_001074397.1 | 6 | 696 | |||
NM_001277116.1 | NP_001264045.1 | 8 | 892 | |||
NM_009035.5 | NP_033061.3 | 6 | 712 | |||
GenBank mRNA | AK028363.1 | - | 8 | 870 | ||
AK080359.1 | - | 6 | 605 | |||
AK090006.1 | - | 6 | 598 | |||
AK139087.1 | - | 6 | 597 | |||
AK145187.1 | - | 6 | 604 | |||
AK152746.1 | - | 6 | 712 | |||
AK153163.1 | - | 7 | 600 | |||
BC035299.1 | - | 4 | 498 | |||
BC051387.1 | - | 6 | 641 | 4460290 | ||
S63463.1 | - | 6 | 713 | |||
X17459.1 | - | 6 | 708 |