Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Relb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290457.1 | NP_001277386.1 | 3 | 315 | ||
NM_009046.2 | NP_033072.2 | 4 | 324 | |||
GenBank mRNA | AK146932.1 | - | 4 | 325 | ||
AK156767.1 | - | 4 | 280 | |||
BC019765.1 | - | 4 | 324 | 3584200 | ||
BC034523.1 | - | 3 | 128 | 4485540 | ||
BC117793.1 | - | 4 | 260 | 40092162 | ||
DQ020178.1 | - | 4 | 134 | |||
DQ020179.1 | - | 3 | 125 | |||
DQ020180.1 | - | 5 | 169 | |||
DQ020181.1 | - | 4 | 134 | |||
M83380.1 | - | 4 | 161 | |||
S56076.1 | - | 4 | 299 |