Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Relb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290457.1 | NP_001277386.1 | 4, 5 | 791 | ||
NM_009046.2 | NP_033072.2 | 5, 6 | 800 | |||
GenBank mRNA | AK146932.1 | - | 5, 6 | 801 | ||
AK156767.1 | - | 5, 6 | 756 | |||
BC019765.1 | - | 5, 6 | 800 | 3584200 | ||
BC034523.1 | - | 4, 5 | 604 | 4485540 | ||
BC117793.1 | - | 5, 6 | 736 | 40092162 | ||
DQ020179.1 | - | 5 | 1568 | |||
DQ020180.1 | - | 7 | 1608 | |||
DQ020181.1 | - | 6 | 1634 | |||
M83380.1 | - | 5, 6 | 637 |