Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271765.1 | NP_001258694.1 | 5 | 1433 | ||
NM_004695.3 | NP_004686.1 | 5 | 1367 | |||
XM_005257790.1 | XP_005257847.1 | 4 | 1309 | |||
XM_011525462.1 | XP_011523764.1 | 4 | 1309 | |||
XM_017025293.1 | XP_016880782.1 | 5 | 1395 | |||
GenBank mRNA | AK000416.1 | - | 5 | 1433 | ||
AK092512.1 | - | 4 | 1476 | |||
AK304163.1 | - | 4 | 1193 | |||
BC009684.1 | - | 5 | 1348 | 3895881 | ||
BC033611.1 | - | 4 | 1257 | 5218712 | ||
U59299.1 | - | 3 | 1121 |