Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009055.4 | NP_033081.3 | 16 | 2337 | ||
GenBank mRNA | AK037226.1 | - | 9 | 1280 | ||
AK039651.1 | - | 2 | 931 | |||
AK083571.1 | - | 16 | 2308 | |||
AK147790.1 | - | 16 | 2342 | |||
BC057018.1 | - | 16 | 2316 | 5705637 | ||
X76088.1 | - | 16 | 2244 |