Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009056.2 | NP_033082.1 | 17 | 2546 | ||
NM_027787.1 | NP_082063.1 | 18 | 2621 | |||
GenBank mRNA | AK044439.1 | - | 18 | 2623 | ||
BC004654.1 | - | 17 | 2563 | 3497910 | ||
X76089.1 | - | 17 | 2498 |