Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145834.1 | NP_001139306.1 | 3 | 627 | ||
NM_001145835.1 | NP_001139307.1 | 3 | 503 | |||
NM_001145836.1 | NP_001139308.1 | 3 | 503 | |||
NM_009058.2 | NP_033084.2 | 3 | 627 | |||
GenBank mRNA | AK129328.1 | - | 3 | 468 | ||
AK141959.1 | - | 3 | 494 | |||
AK159020.1 | - | 3 | 496 | |||
AK159754.1 | - | 3 | 473 | |||
BC027225.1 | - | 3 | 285 | 3486732 | ||
L07924.1 | - | 3 | 463 |