Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009062.3 | NP_033088.2 | 5 | 803 | ||
GenBank mRNA | AK030536.1 | - | 5 | 804 | ||
AK039134.1 | - | 5 | 804 | |||
BC003882.1 | - | 5 | 781 | 3496887 | ||
BC055293.1 | - | 5 | 793 | 6494449 | ||
DQ346659.1 | - | 5 | 803 | |||
DQ346660.1 | - | 5 | 803 | |||
DQ346661.1 | - | 5 | 803 |