Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ring1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009066.3 | NP_033092.3 | 6 | 1294 | ||
GenBank mRNA | AK154962.1 | - | 6 | 1255 | ||
AK155509.1 | - | 6 | 1270 | |||
BC009070.1 | - | 6 | 1257 | 2651194 | ||
BC082771.1 | - | 2 | 2104 | |||
BU505852.1 | - | 1 | 61 | |||
BU918502.1 | - | 2 | 337 | |||
CT010219.1 | - | 4 | 984 | |||
CX219100.1 | - | 1 | 222 | |||
KJ650216.1 | - | 5 | 1071 | |||
Y12881.1 | - | 4 | 990 |