Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277113.1 | NP_001264042.1 | 4 | 1114 | ||
NM_001277114.1 | NP_001264043.1 | 4 | 1184 | |||
NM_009079.3 | NP_033105.1 | 4 | 1164 | |||
GenBank mRNA | AK012339.1 | - | 1 | 298 | ||
AK014372.1 | - | 1 | 298 | |||
AK169078.1 | - | 4 | 1017 | |||
AK169731.1 | - | 4 | 1030 | |||
BC021344.1 | - | 4 | 1023 | 4190569 |