Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009082.2 | NP_033108.1 | 4 | 420 | ||
GenBank mRNA | AK013307.1 | - | 4 | 420 | ||
AK088551.1 | - | 4 | 359 | |||
AK141477.1 | - | 4 | 385 | |||
AK167284.1 | - | 4 | 362 | |||
BC002062.1 | - | 338 | 3590425 | |||
BC019179.1 | - | 349 | ||||
BC081467.1 | - | 3 | 332 | 6823037 | ||
BC082292.1 | - | 4 | 338 | 6487045 | ||
BC086897.1 | - | 4 | 366 | 6775819 | ||
BC086898.1 | - | 4 | 362 | 30120979 | ||
BC087950.1 | - | 4 | 362 | 30122718 | ||
BC092262.1 | - | 4 | 348 | 2651191 | ||
BC094039.1 | - | 4 | 348 | 5006561 | ||
BC096575.1 | - | 4 | 355 | 1430129 | ||
L08651.1 | - | 4 | 338 |
Gene Symbol: Gm8580 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CJ081075.1 | - | 1 | 98 | ||
CJ084848.1 | - | 1 | 98 |