Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009090.5 | NP_033116.2 | 6 | 516 | ||
GenBank mRNA | AK137796.1 | - | 449 | |||
AK146763.1 | - | 6 | 446 | |||
AK151270.1 | - | 6 | 444 | |||
AK160053.1 | - | 6 | 451 | |||
BC002023.1 | - | 6 | 434 | 3492868 | ||
BC099548.1 | - | 6 | 461 | 30312083 | ||
D83999.1 | - | 6 | 437 |