Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009099.2 | NP_033125.2 | 5, 6 | 1006 | ||
GenBank mRNA | AF220014.1 | - | 5, 6 | 923 | ||
AK134144.1 | - | 5, 6 | 975 | |||
AK137506.1 | - | 5, 6 | 1007 | |||
BC005447.1 | - | 5, 6 | 940 | 3493256 | ||
J03776.1 | - | 5, 6 | 923 |