Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sap18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009119.3 | NP_033145.2 | 4 | 877 | ||
GenBank mRNA | AK085992.1 | - | 4 | 816 | ||
AK133619.1 | - | 4 | 811 | |||
BC006625.1 | - | 4 | 818 | 3968600 | ||
Z97062.1 | - | 4 | 812 |