Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291865.1 | NP_001278794.1 | 7 | 767 | ||
NM_009121.4 | NP_033147.1 | 6 | 657 | |||
GenBank mRNA | AK002531.1 | - | 6 | 558 | ||
AK131657.1 | - | 3 | 944 | |||
AK154445.1 | - | 6 | 553 | |||
BC058696.1 | - | 6 | 617 | 6820169 | ||
BC108334.1 | - | 6 | 968 | |||
L10244.1 | - | 6 | 465 |