Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009140.2 | NP_033166.1 | 4 | 859 | ||
GenBank mRNA | AK137628.1 | - | 4 | 887 | ||
AK150450.1 | - | 4 | 767 | |||
AK155458.1 | - | 1 | 1843 | |||
AK155874.1 | - | 3 | 1637 | |||
AK155916.1 | - | 4 | 886 | |||
AK157079.1 | - | 4 | 886 | |||
X53798.1 | - | 4 | 859 |