Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293673.1 | NP_001280602.1 | 1, 2 | 294 | ||
NM_009145.2 | NP_033171.2 | 1, 2 | 294 | |||
GenBank mRNA | AK075610.1 | - | 1, 2 | 129 | ||
AK153930.1 | - | 1, 2 | 127 | |||
BC039191.1 | - | 1, 2 | 144 | |||
D50463.1 | - | 1, 2 | 125 |