Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009169.2 | NP_033195.1 | 3 | 392 | ||
GenBank mRNA | AK135462.1 | - | 345 | |||
AK160432.1 | - | 3 | 385 | |||
AK161690.1 | - | 3 | 365 | |||
AK167914.1 | - | 3 | 342 | |||
BC023490.1 | - | 3 | 349 | 5251089 | ||
BC058117.1 | - | 3 | 358 | 6412144 | ||
U41626.1 | - | 3 | 334 | |||
X83589.1 | - | 3 | 367 |