Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009170.3 | NP_033196.1 | 3 | 1988 | ||
GenBank mRNA | AK077688.1 | - | 3 | 1989 | ||
BC063087.1 | - | 3 | 1977 | 6516263 |