Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042576.1 | NP_001036041.1 | 3 | 788 | ||
NM_004587.2 | NP_004578.2 | 2 | 711 | |||
GenBank mRNA | AB037819.2 | - | 718 | |||
AB287347.2 | - | 1 | 434 | |||
AF006751.1 | - | 2 | 504 | |||
BC009700.1 | - | 3 | 647 | |||
BC108741.1 | - | 2 | 514 |