Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6galnac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009180.3 | NP_033206.2 | 4 | 547 | ||
GenBank mRNA | AK004613.1 | - | 4 | 548 | ||
AK136381.1 | - | 550 | ||||
BC003276.1 | - | 4 | 1449 | |||
BC010208.1 | - | 499 | ||||
X93999.1 | - | 4 | 511 |