Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009181.2 | NP_033207.2 | 3 | 392 | ||
GenBank mRNA | BC096546.1 | - | 3 | 384 | 6833487 | |
X83562.1 | - | 3 | 371 |