Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009182.3 | NP_033208.2 | 6 | 2043 | ||
GenBank mRNA | AK133780.1 | - | 4 | 1328 | ||
AK134678.1 | - | 4 | 1339 | |||
AK161482.1 | - | 6 | 2058 | |||
BC075645.1 | - | 4 | 1338 | 30620472 | ||
X80502.1 | - | 4 | 1328 |