Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7a2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159529.1 | NP_001153001.1 | 2 | 214 | ||
NM_009187.3 | NP_033213.1 | 2 | 214 | |||
GenBank mRNA | AK009614.1 | - | 2 | 211 | ||
AK133520.1 | - | 2 | 214 | |||
AK150034.1 | - | 2 | 211 | |||
AK152553.1 | - | 2 | 204 | |||
AK159441.1 | - | 2 | 212 | |||
AK166761.1 | - | 2 | 199 | |||
AK167062.1 | - | 2 | 209 | |||
AK169619.1 | - | 2 | 226 | |||
AK184659.1 | - | 1 | 89 | |||
AK216072.1 | - | 1 | 89 | |||
AK216357.1 | - | 1 | 89 | |||
BC004715.1 | - | 2 | 178 | 3582142 | ||
BC085296.1 | - | 2 | 183 | 5337317 | ||
BY122624.1 | - | 2 | 214 | |||
X80899.1 | - | 2 | 182 |