Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009190.2 | NP_033216.2 | 7 | 1033 | ||
GenBank mRNA | AK151023.1 | - | 7 | 987 | ||
AK152131.1 | - | 7 | 1020 | |||
AK165594.1 | - | 978 | ||||
AK168270.1 | - | 7 | 924 | |||
BC003799.1 | - | 7 | 940 | 3488471 | ||
U10119.1 | - | 7 | 984 |