Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009190.2 | NP_033216.2 | 11 | 2757 | ||
GenBank mRNA | AK034641.1 | - | 1415 | |||
AK151023.1 | - | 11 | 2711 | |||
AK152131.1 | - | 11 | 2744 | |||
AK168270.1 | - | 11 | 2648 | |||
BC003799.1 | - | 11 | 2664 | 3488471 | ||
U10119.1 | - | 11 | 2699 |