Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009191.3 | NP_033217.1 | 5 | 729 | ||
GenBank mRNA | AK006072.1 | - | 6 | 994 | ||
AK028101.1 | - | 5 | 738 | |||
AK032421.1 | - | 724 | ||||
AK138593.1 | - | 5 | 767 | |||
AK141383.1 | - | 5 | 738 | |||
AK145641.1 | - | 5 | 721 | |||
AK147772.1 | - | 5 | 450 | |||
AK154577.1 | - | 5 | 750 | |||
AK159314.1 | - | 5 | 770 | |||
BC048175.1 | - | 5 | 729 | 6825472 | ||
U09874.1 | - | 5 | 731 |