Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243879.1 | NP_001230808.1 | 8 | 1311 | ||
NM_004593.2 | NP_004584.1 | 9 | 1445 | |||
XM_005247703.1 | XP_005247760.1 | 10 | 1613 | |||
XM_011513072.2 | XP_011511374.1 | 10 | 4415 | |||
XM_017007033.1 | XP_016862522.1 | 9 | 2591 | |||
GenBank mRNA | AB209208.1 | - | 10 | 4401 | ||
AK091863.1 | - | 5 | 2332 | |||
AK094495.1 | - | 6 | 1367 | |||
AK098191.1 | - | 9 | 1232 | |||
AK125610.1 | - | 6 | 2115 | |||
BC000160.2 | - | 9 | 1312 | 2984771 | ||
BC000451.2 | - | 9 | 1311 | 2821443 | ||
BC005898.1 | - | 9 | 1296 | 4046349 | ||
CR749214.1 | - | 8 | 3472 | |||
U61267.1 | - | 9 | 1290 | |||
U68063.1 | - | 9 | 1298 |