Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253804.1 | NP_001240733.1 | 2 | 279 | ||
NM_009195.3 | NP_033221.1 | 2 | 273 | |||
GenBank mRNA | AF047339.1 | - | 2 | 197 | ||
AF121118.1 | - | 2 | 264 | |||
AK149969.1 | - | 2 | 262 | |||
AK151717.1 | - | 2 | 234 | |||
AK152762.1 | - | 2 | 251 | |||
AK153245.1 | - | 2 | 251 | |||
AK159274.1 | - | 2 | 261 | |||
AK159331.1 | - | 2 | 256 | |||
AK159483.1 | - | 2 | 229 | |||
AK159537.1 | - | 2 | 256 | |||
AK160034.1 | - | 2 | 222 | |||
AK160072.1 | - | 2 | 256 | |||
AK164479.1 | - | 2 | 238 | |||
AK165874.1 | - | 2 | 251 | |||
AK167371.1 | - | 2 | 270 | |||
AK167444.1 | - | 2 | 270 | |||
AK169044.1 | - | 2 | 228 | |||
BC066872.1 | - | 2 | 203 | 30436525 | ||
BY740705.1 | - | 2 | 274 |