Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253804.1 | NP_001240733.1 | 18, 19 | 2519 | ||
NM_009195.3 | NP_033221.1 | 18, 19 | 2513 | |||
GenBank mRNA | AF047339.1 | - | 18, 19 | 2437 | ||
AF121118.1 | - | 18, 19 | 2504 | |||
AK149969.1 | - | 18, 19 | 2502 | |||
AK151717.1 | - | 18, 19 | 2474 | |||
AK152762.1 | - | 18, 19 | 2491 | |||
AK153245.1 | - | 18, 19 | 2491 | |||
AK159274.1 | - | 18, 19 | 2501 | |||
AK159331.1 | - | 18, 19 | 2496 | |||
AK159483.1 | - | 18, 19 | 2469 | |||
AK159537.1 | - | 18, 19 | 2496 | |||
AK160034.1 | - | 18, 19 | 2462 | |||
AK160072.1 | - | 18, 19 | 2496 | |||
AK164479.1 | - | 18, 19 | 2478 | |||
AK165874.1 | - | 18, 19 | 2491 | |||
AK166304.1 | - | 18, 19 | 2472 | |||
AK167371.1 | - | 18, 19 | 2510 | |||
AK167444.1 | - | 18, 19 | 2510 | |||
AK169044.1 | - | 18, 19 | 2452 | |||
AK185898.1 | - | 1, 2 | 22 | |||
AK193687.1 | - | 1, 2 | 22 | |||
AK200839.1 | - | 1, 2 | 22 | |||
BC066872.1 | - | 18, 19 | 2443 | 30436525 |