Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170638.1 | NP_001164109.1 | 5 | 621 | ||
NM_009198.3 | NP_033224.3 | 5 | 713 | |||
GenBank mRNA | AA986929.1 | - | 3 | 380 | ||
AK132800.1 | - | 5 | 1262 | |||
BC013445.1 | - | 5 | 654 | 4241557 | ||
CJ065843.1 | - | 5 | 622 | |||
X77241.1 | - | 5 | 580 |