Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009205.2 | NP_033231.2 | 1 | 448 | ||
GenBank mRNA | AB009290.1 | - | 1 | 433 | ||
AK052623.1 | - | 1 | 419 | |||
AK132275.1 | - | 1 | 420 | |||
BC013441.1 | - | 1 | 390 | 4222972 | ||
CA493293.1 | - | 1 | 493 | |||
D88533.1 | - | 1 | 408 |