Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009205.2 | NP_033231.2 | 3 | 779 | ||
GenBank mRNA | AK052623.1 | - | 3 | 750 | ||
AK132275.1 | - | 3 | 751 | |||
BC013441.1 | - | 3 | 721 | 4222972 | ||
D88533.1 | - | 3 | 739 |