Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009211.2 | NP_033237.2 | 16 | 1601 | ||
GenBank mRNA | AK143485.1 | - | 16 | 1589 | ||
AK144122.1 | - | 16 | 1575 | |||
AK144631.1 | - | 16 | 1569 | |||
AK145352.1 | - | 16 | 1568 | |||
AK204398.1 | - | 3 | 189 | |||
BC022173.1 | - | 16 | 1565 | |||
BC052423.1 | - | 16 | 1563 | 6400646 | ||
BC053064.1 | - | 16 | 1557 | |||
BC056375.1 | - | 16 | 1564 | |||
U85614.1 | - | 16 | 1562 |