Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009229.4 | NP_033255.1 | 2 | 728 | ||
GenBank mRNA | AK040423.1 | - | 2 | 729 | ||
AK080059.1 | - | 2 | 725 | |||
AK131687.1 | - | 2 | 753 | |||
AK165009.1 | - | 2 | 730 | |||
AK165900.1 | - | 2 | 705 | |||
BC138850.1 | - | 2 | 727 | 8861872 | ||
BC145442.1 | - | 2 | 727 | 9054000 | ||
U00678.1 | - | 2 | 727 |