Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009229.4 | NP_033255.1 | 7 | 2910 | ||
GenBank mRNA | AK010705.1 | - | 1 | 322 | ||
AK040423.1 | - | 7 | 2911 | |||
AK080059.1 | - | 7 | 2907 | |||
BC085295.1 | - | 1 | 409 | |||
BC138850.1 | - | 7 | 2909 | 8861872 | ||
BC145442.1 | - | 6 | 2724 | 9054000 | ||
U00678.1 | - | 7 | 2908 |