Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009231.2 | NP_033257.2 | 3, 4 | 882 | ||
GenBank mRNA | AK140310.1 | - | 3, 4 | 882 |