Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004616.2 | NP_004607.1 | 9 | 1075 | ||
XM_006719583.3 | XP_006719646.1 | 9 | 1029 | |||
XM_017019913.1 | XP_016875402.1 | 8 | 1111 | |||
GenBank mRNA | AK223022.1 | - | 8 | 1003 | ||
BC005246.1 | - | 9 | 1075 | 3934330 | ||
BC070168.1 | - | 13 | 1449 | |||
M35252.1 | - | 8 | 1033 |