Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204201.1 | NP_001191130.1 | 7 | 525 | ||
NM_001204202.1 | NP_001191131.1 | 6 | 522 | |||
NM_001204203.1 | NP_001191132.1 | 7 | 473 | |||
NM_001204233.1 | NP_001191162.1 | 5 | 567 | |||
NM_009263.3 | NP_033289.2 | 6 | 470 | |||
GenBank mRNA | AF515708.1 | - | 5 | 370 | ||
AK133555.1 | - | 6 | 461 | |||
AK149588.1 | - | 6 | 461 | |||
AK150819.1 | - | 6 | 461 | |||
AK150845.1 | - | 6 | 461 | |||
AK151069.1 | - | 6 | 431 | |||
AK151562.1 | - | 6 | 461 | |||
AK153089.1 | - | 6 | 460 | |||
AK154908.1 | - | 6 | 461 | |||
AK160540.1 | - | 6 | 464 | |||
AK165393.1 | - | 6 | 461 | |||
AK165965.1 | - | 6 | 513 | |||
AK171158.1 | - | 6 | 461 | |||
BC057858.1 | - | 6 | 443 | 5294167 | ||
CT010357.1 | - | 5 | 370 | |||
J04806.1 | - | 6 | 439 | |||
X13986.1 | - | 5 | 385 | |||
X16151.1 | - | 5 | 592 |