Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sephs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009266.3 | NP_033292.2 | 1 | 696 | ||
GenBank mRNA | AK077573.1 | - | 1 | 697 | ||
AK149516.1 | - | 1 | 693 | |||
AK168722.1 | - | 1 | 697 | |||
AK168965.1 | - | 1 | 697 | |||
BC003760.1 | - | 1 | 683 | |||
BC016508.1 | - | 1 | 688 | 4911695 | ||
BC028966.1 | - | 1 | 673 | 5374184 | ||
BC043334.1 | - | 1 | 677 | 5100207 | ||
U43285.1 | - | 1 | 693 |